VWR® SAS Super Isolator Air Samplers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710-0943EA 3830 GBP
710-0943 710-0944 710-0977 710-0978 710-0945 710-0946
VWR® SAS Super Isolator Air Samplers
Samplers Air Samplers
The SAS instrument control unit is placed outside and connected to the aspirating head using a simple electrical connection. All possible typical contamination sources from the vacuum sampling system are avoided using the SAS Super Isolator. An interesting application is that several heads can be controlled by a single SAS control unit, providing a practical and economical solution.
  • Operational flexibility, multiple language choice
  • Records day, time, site, volume of air sampled and operator's name
  • Large backlit screen with intuitive menu; active operation
More Product Information