Total Organic Carbon (TOC) cleaning validation sets
Supplier: Texwipe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TX3340 TX3342 TX3350 TX3343
710-0542EA 200 GBP
710-0542 710-0582 115-2152 115-2149
Total Organic Carbon (TOC) cleaning validation sets
Swabs Swabs for Cleaning
These sets contain components for sampling different locations in the production area, and provide an efficient means of transporting the samples from production to laboratory with minimal chance of contamination.
  • Low particle generation breakaway handle allows swab head to be placed into the vial with minimal handling and contamination
  • Good absorbent capacity, excellent chemical resistance
  • Autoclavable to ensure sampling under sterile conditions
Designed to simplify sampling for cleaning validation in the pharmaceutical industry.
Certifications: More Product Information