Colour standards
Supplier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
141453 141410 2602853
707-0002EA 394 GBP
707-0002 707-0001 707-0009
Colour standards
Standards Colour Reference and Spectroscopy Standards
For accuracy checks in colour determinations.
  • Simple Accuracy Checks
  • Minimize waste with Voluette ampoules
  • Reliable results
The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.