Bench top turbidity meters, TL23
Supplier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LPV4449900210 LPV4449900120 LPV4449900310 LPV44499.00320
705-0982EA 5150 GBP
705-0982 705-0983 705-0984 705-1176
Bench top turbidity meters, TL23
Turbidity Meters
Tungsten or LED light sources, as well as proven optics, make these instruments more reliable to deliver accurate and stable measurements.
  • Improved and intuitive design with guided calibration and verification
  • USB port for data export and sample identification
  • Ratio technology that helps to overcome interference from colour and large particles
  • Coloured tactile LCD with commands
  • 90° detector
Designed for simple use in demanding applications of pharmaceutical, cosmetic, food, chemical and petrochemical industries.
Certifications: US EPA 180.1 or ISO 7More Product Information