Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

7033.1000EA 0 GBP
7033.1000 7033.5000JC 7033.9200
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min. 99.0 %
Color (APHA) max. 10
Density (g/ml) at 25°C min. 1.095
Freezing Point 18.0 - 19.0 °C
Residue after Evaporation max. 0.01 %
Water (H2O) max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR