Diethanolamine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

7026-00EA 0 GBP
7026-00 7026-01RM
Diethanolamine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Diethanolamine
Formula: C₄H₁₁NO₂
Boiling Pt: 268.8 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Specification Test Results

Assay min. 98.5 %
Density (g/ml) at 30°C 1.088 - 1.093
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.002 %
Residue after Ignition max. 0.005 %
Trace Impurities (in ppm) Chloride (Cl) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR