Water samplers, SP 5 series
Supplier: MAXX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030012 0030013 0060144 0060342 0060161 0060045 0060036 0060035 0060034 V1300001 1200000 V1200012 V1200024 V1200001 V1200002 V1200004 V1200012G V1200024G V1200040 V1200050 V1200414 V1300012 V1300024 V1300024G V1300012G V1300414
701-0098EA 78.1 GBP
701-0098 701-0099 701-0100 701-0105 701-0101 701-0075 701-0102 701-0104 701-0103 701-0150 701-0139 701-0144 701-0146 701-0140 701-0141 701-0142 701-0143 701-0145 701-0147 701-0148 701-0149 701-0152 701-0154 701-0153 701-0151 701-0155
Water samplers, SP 5 series
Samplers Water Samplers
The SP 5 are modular fixed site units available in different configurations.
  • Thermostatic control of the sample compartment to 4 °C with overheating protection and auto defrost
  • Microprocessor to control and select sampling and bottle filling intervals with real time events
  • Diaphragm pump with metering vessel made of glass Duran 50
Adjustable accurate sample volumes for collection into the single or multiple containers.