N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12), Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: sulphobetaine-12, 3-(Dodecyldimethylammonio)propane-1-sulphonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulphonate

693015-25 693015-5 693015-1KG
693015-25EA 420 GBP
693015-25 693015-5 693015-1KG
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12), Millipore®
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-12, SB-12)
Formula: C₁₇H₃₇NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036909
CAS Number: 14933-08-5
EINECS: 239-002-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR