Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

673811-4L 673811-4X4L
673811-4LEA 546 GBP
673811-4L 673811-4X4L
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Diethyl ether

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602

Formula: (CH₃CH₂)₂O
Boiling Pt: 34…35 °C (1.013 hPa)
Melting Pt: –116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: –45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Titrable Acid Max. 0.0002 meq/g
Peroxides (as H₂O₂) Max. 1 ppm
Ethanol (GC) Max. 0.05 %
Carbonyl Compounds (as HCHO) Max. 0.001 %
Assay BHT (GC) Min. 1 ppm
APHA Max. 10
Vapor Pressure 8.56 psi ( 20 °C)
Refractive Index n20/D 1.3530 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR