Methanol, anhydrous ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

6712-25EA 43.7 GBP
6712-25
Methanol, anhydrous ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Methanol
Formula: CH₃OH
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) ≥ 99.8 %
Water (KF) ≤ 0.05 %
Titrable Acid (meq/g) ≤ 0.0005
Titrable Base (meq/g) ≤ 0.0005
Residue after Evaporation ≤ 0.0005 %
Color (APHA) ≤ 10
Ultraviolet Absorbance (1.00-cm cell vs. water): 0
210 nm ≤ 0.6
220 nm ≤ 0.3
230 nm ≤ 0.2
240 nm ≤ 0.05
260 nm ≤ 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR