VWR® Silicone Fluids, Silicone Oils 47
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6678.1000EA 240 GBP
6678.1000 83851.290 83851.360
VWR® Silicone Fluids, Silicone Oils 47
Circulator Bath Fluids
Silicone Oils 47 are polydimethylsiloxane oils, consisting of linear molecular chains of varying lengths whose groups comprise alternating silicone and oxygen atoms (the Si-O-Si siloxane bond). The silicone atoms are saturated by methyl groups – CH₃.
  • Heat transfer and dielectric fluids
  • Demolding or release agents
  • Foam control agents
  • Lubricants
  • Active components in maintenance product formulations
The oils display low and high temperature resistance, resistance to oxidation and hydrolysis, lack of ageing by atmospheric agents (oxygen-ozone-water-light-UV) and are chemically inert (no risk of corrosion).