Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9620 IDS
Supplier: WTW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1FD560 1FD56C
665-0476EA 2530 GBP
665-0476 665-0560
Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9620 IDS
Electrochemistry Meters pH and Multi-Parameter Meters
Digital high performance multi-parameter device with two galvanically isolated measuring channels for pH/mV, ISE (including incremental methods), conductivity and oxygen measurements. With comprehensive GLP functions.
  • Simultaneous measurement with two independent measuring channels
  • User-defined combinations of equal or different parameters
  • Continuous Measurement Control (CMC) for measuring in the optimum pH range
  • Quality Sensor Control (QSC) benchmarks the current condition of the special IDS electrodes
  • Digital processing and data transfer (GLP compliant)
More Product Information