Multi-parameter meters, handheld, 3320
Supplier: WTW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2FA310 2FA311 2FA312 2EA310 2EA312
665-0319EA 1840 GBP
665-0319 665-0320 665-0321 665-0313 665-0315
Multi-parameter meters, handheld, 3320
Electrochemistry Meters pH and Multi-Parameter Meters
Multi 3320: This advanced meter features two inputs for the simultaneous measurement of pH and conductivity, or pH and dissolved oxygen. It is perfect for measurements in ground and surface water, aquacultures and wastewater.
  • Backlit LCD for displaying two measurements at the same time
  • Robust and waterproof housing (IP 67)
  • Memory with data logging functionality between 1 s and 60 min
  • All entries are GLP compliant with time, date and selectable ID number
  • Waterproof USB interface enables fast and easy data transfer
pH/Cond 3320: This portable meterMore Product Information