inoLab® Multi 9310 IDS Multi-Parameter Meter (pH/Conductivity/DO), Bench
Supplier: WTW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1FD350P 1FD350 1FD351 1FD352 1FD353 1FD354 1FD35C 1FD35CP
665-0248EA 2090 GBP
665-0248 665-0247 665-0249 665-0251 665-0253 665-0255 665-0257 665-0258
inoLab® Multi 9310 IDS Multi-Parameter Meter (pH/Conductivity/DO), Bench
Electrochemistry Meters pH and Multi-Parameter Meters
The inoLab® Multi 9310 is ideal for all laboratories that require complete documentation, or want to have the most efficient, error-free measurement processes. For the measurement of a single parameter from pH, conductivity and DO.
  • Continuous Measurement Control (CMC) for measuring in the optimum pH range
  • Digital sensor recognition
  • Quality Sensor Control (QSC) benchmarks the current condition of the special IDS pH electrodes
  • Digital processing and data transfer (GLP compliant) via USB interface
  • Optional built-in printer for printing of all measurement and
More Product Information