Multi-parameter meter, handheld, Professional Plus
Supplier: YSI ENVIRONMENTAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
605101 605203 605202 605102 605790-1 605790-4 605790-10 605790-20 605790-30 605104 605105 605106
665-0211EA 477 GBP
665-0211 665-0210 665-0209 665-0208 665-0203 665-0207 665-0204 665-0205 665-0206 665-0212 665-0213 665-0214
Multi-parameter meter, handheld, Professional Plus
Electrochemistry Meters pH and Multi-Parameter Meters
The Professional Plus is one of the most versatile multi-parameter handheld meters on the market. A combination of up to 11 parameters can be measured on a single bulkhead, including pH, ORP, ammonia, nitrate, chloride, DO, conductivity, salinity, TDS, barometric pressure and temperature, meaning only one cable is required.
  • Rubber, over-moulded IP 67 housing (even without battery cover)
  • Rugged cable sensors with military specification, waterproof connectors
  • USB connection and Data Manager software with GLP functions
  • 5000 data sets memory
More Product Information