Oxygen Sensor Agile, Memosens COL37E
Supplier: Endress+Hauser
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
COL37E-1020/0
664-0242EA 1310 GBP
664-0242
Oxygen Sensor Agile, Memosens COL37E
Electrodes Dissolved Oxygen Electrodes
Optical oxygen sensor with stainless steel for laboratory measurements and random sampling in the field. Connectable via inductive, digital connection with Memosens technology.
  • C-shaped optical cap for stable measurements without interference from air bubbles
  • Excellent performance and low maintenance due to optical oxygen measuring principle
The optical Memosens 2.0 oxygen sensor COL37E can be used in all environments and enables a quick check of oxygen levels in aeration tanks, fish farms and surface water.
Thanks to Memosens technology, theMore Product Information