Dissolved oxygen meter, handheld, FiveGo™ F4
Supplier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30266884 30266885 30266886
664-0213EA 360 GBP
664-0213 664-0214 664-0215
Dissolved oxygen meter, handheld, FiveGo™ F4
Electrochemistry Meters Oxygen Meters
These portable meters provide high quality dissolved oxygen measurements with the simple click of a button. The robust and waterproof design makes the FiveGo instruments the perfect companion to get reliable measurement data even in harsh conditions. Ideal for mobile applications in the laboratory, at-line, or outdoors.
  • Large, well structured display, intuitive button arrangement and simple menu
  • Ergonomic design makes it comfortable to work with
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • IP 67 waterproof protection enables
More Product Information