pH buffers, ready to use

Supplier: WTW
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

108706 108708 108800 108802 108805 108703 108698 108700 108702 108722 108825 108811 108812 108824 108827 108801 108803 108809 108814 108826
662-0319EA 22.9 GBP
662-0319 662-0320 662-8222 662-8224 WTWA108805 662-0270 662-0329 662-8232 662-8234 WTWA108722 WTWA108825 WTWA108811 WTWA108812 WTWA108824 WTWA108827 WTWA108801 WTWA108803 WTWA108809 WTWA108814 WTWA108826
pH buffers, ready to use
Buffers pH Calibration Buffers

Every bottle has its own lot reference with the expiration date.


  • Shelf life up to 3 years (unopened)
  • Reference temperature at 25 °C
  • Economic consumption
  • Easiest handling by use of the dosing vessel
  • One litre container with a separate compartment for the sample


Accuracy is ±0,03 pH


Certifications: Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR