Combined ion-selective electrodes, perfectION™
Supplier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51344706 51344727 51344730 51344724 51344721 51344718 51344715 51344712 51344709 51344703 51344700 51344830 51344827 51344821 51344818 51344815 51344812 51344809 51344806 51344800 51344803
662-1280EA 1185 GBP
662-1280 662-1870 662-1914 662-1282 662-1287 662-1286 662-1281 662-1913 662-1912 662-1297 662-1911 662-1929 662-1928 662-1919 662-1918 662-1284 662-1917 662-1916 662-1283 662-1915 662-1298
Combined ion-selective electrodes, perfectION™
Electrodes ISE Electrodes
Combined ion-selective electrodes consisting of a measuring electrode complete with an integrated reference.
  • Click & Clear™ junction enables optimum contact between the electrolyte and measurement solution, even in difficult samples
  • The electrolyte solution can be made to flow easily through the junction by pressing the electrode head downwards
  • The junction needs to be cleaned between measurements and the electrolyte can be simply emptied and refilled if necessary
Other electrodes available on request. For further information please contact yourMore Product Information