pH/mV/°C meters, handheld, Orion™ Star A121
Supplier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
STARA1210 STARA1215
662-1440EA 434 GBP
662-1440 662-1441
pH/mV/°C meters, handheld, Orion™ Star A121
Electrochemistry Meters pH and Multi-Parameter Meters
Easy-to-use and budget-friendly instrument for basic pH analysis in the field. Ideal for simple, routine pH, mV and temperature measurements.
  • Up to a three point pH calibration with automatic buffer recognition or manual buffer entry
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
  • User-friendly operation with basic calibration text prompts
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss
More Product Information