pH combination electrodes with temperature sensor, ScienceLine
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285100486 285104541 285130200 285100510 285093009 285101290 285130275
662-6111EA 444 GBP
662-6111 662-6106 662-1972 662-1949 662-1941 662-1377 661-0243
pH combination electrodes with temperature sensor, ScienceLine
Electrodes pH Electrodes
Electrodes with plug head (without cable) or with BNC/DIN plug with fixed cable.
  • Reference system: Silamid®
  • Membrane shape: sphere
  • Electrodes with suffix 'ID' feature an ID function (electrode data is stored in the connector for the automatic sensor recognition)