Combined ion selective electrodes
Supplier: SENTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
360-75 361-75 366-75 362-75BNC 367-75 368-75 370-75 371-75 365-75BNC 379-75 380-75 372-75 373-75 364-75BNC 375-75 376-75 377-75 315-75BNC 321-75 378-75
662-1181EA 3320 GBP
662-1181 662-1182 662-1186 662-1183 662-1187 662-1188 662-1189 662-1190 662-1185 662-1197 662-1198 662-1191 662-1192 662-1184 662-1193 662-1194 662-1195 662-1179 662-1180 662-1196
Combined ion selective electrodes
Electrodes ISE Electrodes
Rugged solid-state electrodes with built-in reference elements that do not require any filling solution or maintenance. Ideal for unskilled operatives.
  • Fully submersible and waterproof
  • Virtually unbreakable
  • Colour coded caps for identification in the field
  • Can be left dry for long periods
  • Long lifetime
  • With 1 m cable
Order Now
SPECIFICATIONS
Environmentally Preferable