pH electrode, micro, Orion™ 9863BN
Supplier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9863BN
662-0889EA 1790 GBP
662-0889
pH electrode, micro, Orion™ 9863BN
Electrodes pH Electrodes
This bevel needle-tipped 16-gauge glass body combination electrode can be used to measure pH by piercing septa.
  • Made of very thin, delicate glass
  • Single junction Ag/AgCl
  • Refillable