Conductivity cells
Supplier: WTW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
301602 301960 301961 301962 301970 301971 301972 301973 301974 301900 301995
662-0481EA 1120 GBP
662-0481 663-8117 663-8118 662-0484 663-1841 663-1842 663-1843 663-1844 663-1845 663-0227 663-0146
Conductivity cells
Electrodes Conductivity Sensors
TetraCon® 325, TetraCon® V and KLE 325 are suitable for on-site measurements in rivers, lakes and wastewater, fish farming, ground water measurements and applications in water laboratories.
TetraCon® 325/C is ideal for heavy chemical applications, e.g. for acid media.
TetraCon® 325/S is great for measurements in pastes, creams and emulsions.
LR 325/01 is good for measurements in ultapure water.