Titrators, potentiometric and volumetric, Titroline® 7750
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285220250 285220240
661-1007EA 5960 GBP
661-1007 661-1006
Titrators, potentiometric and volumetric, Titroline® 7750
Titrators
The Titroline® 7750 is the all-rounder for both potentiometric titration and volumetric KF titration. The titrator combines the features of the potentiometric titrator TitroLine® 7000 and the volumetric Karl Fischer titrator TitroLine® 7500.
  • Highly visible full colour display, that can be easily viewed from a distance and extreme angles
  • With standard methods for potentiometric and KF titration and interchangeable modules (5, 10, 20 and 50 ml)
  • System is easily expendable thanks to the USB and RS232 hosts
  • Data storage of results (PDF and CSV format)
More Product Information