Inspection sets, PosiTector®, DeFelsko®

Supplier: Industrial Physics
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

LD6003 LD6004 LD6005 LD6006 LD6001 LD6008 LD6007 LD6002
649-0028EA 2460 GBP
649-0028 649-0029 649-0030 649-0031 649-0026 649-0033 649-0032 649-0027
Inspection sets, PosiTector®, DeFelsko®
Coating Testers
Electronic coating gauges use magnetic and eddy current principles to measure coating thickness on both ferrous and non-ferrous metals with all PosiTector 6000, 200, DPM, UTG, RTR and SPG probes.

  • Solvent, acid, oil, water and dust resistant, weatherproof
  • Environmental probe measurements and records of climatic conditions
  • Surface profile probe can measure peak to valley surface profile height
  • Storage of 250 (Standard) or 100 000 readings (Advanced)
  • Easily converting from a coating thickness gauge to a dew point meter, surface profile gauge or wall thickness
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR