VWR®, Bench Top Refractometers with Peltier Temperature Control, PLR1/PLR2
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
635-0844EA 9360 GBP
635-0844 635-0845
VWR®, Bench Top Refractometers with Peltier Temperature Control, PLR1/PLR2
Refractometers Benchtop Refractometers
Simple to use, precise bench top refractometers for use in laboratory or production environments.
  • Wide measuring range
  • Brix, RI, user scales
  • Peltier temperature control
  • 7" touch screen
  • Multilingual display
  • Intuitive menu structure
  • Store up to 4000 readings
Two distinct high precision bench top refractometers that offer users a choice based on measurement range and application type. Both models feature refractive index (nD) and sugar (Brix) scales and it is possible to load up to 25 commonly used scales from the instrument’s library.
The PLR1 isMore Product Information