Digital polarimeter, AP-300
Supplier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5291 RE-72043 RE-72044 RE-72045
635-0409EA 8250 GBP
635-0409 635-0500 635-0501 635-0502
Digital polarimeter, AP-300
Polarimeters
AP-300 with automatic temperature compensation (ATC) for sugar factories.
  • Password function to identify operators and protect data
  • Built-in International Sugar Scale (ISS)
  • Manual calibration with quartz control plates
Temperature control is not necessary for users who have temperature controlled working environments or are able to benefit from the temperature compensation funtionality.
The Pharmaceutical packages include a GMP and GLP compliant quartz control plate for calibration and a digital printer DP-AD (regular paper) for data recording.