Digital pocket refractometers for food, PAL series
Supplier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4427 4429 4430 4439 4498
635-0234EA 323 GBP
635-0234 635-0235 635-0236 635-0243 ATAG4498
Digital pocket refractometers for food, PAL series
Refractometers Handheld Refractometers
Battery operated instruments for food applications.
  • Single handed operation
  • Easy to use and delivering rapid results
  • Corresponding temperature range also displayed
Certifications: IP 65 for rinsing off the sample with running water