VWR®, Disposable Cuvettes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
634-0678EA 19.8 GBP
634-0678 634-0675 634-0676 634-0677
VWR®, Disposable Cuvettes
Cuvettes Spectrophotometer Cuvettes
Disposable cuvettes in either optical polystyrene (PS) or polymethyl methacrylate (PMMA), for use with most polar solvents, acids and alkaline solutions.
  • Minimum variation of extinction values
  • Recessed windows, clear and clean optical path
  • Optimum transmittance and reduced risk of contamination
For determinations in water analysis, chemistry and life science applications.
Packaging: Resealable – cuvettes are assorted by cavity numbers.