Nessleriser systems, Lovibond® comparators with Pt-Co/hazen/APHA scale
Supplier: Lovibond Tintometer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
433290 433270
633-0150EA 2230 GBP
633-0150 633-0149
Nessleriser systems, Lovibond® comparators with Pt-Co/hazen/APHA scale
Colorimeters Comparator Kits
For colour grading of clear, light yellow liquids such as clear oils, chemicals, petrochemicals and water. In particular, the Nessleriser 1209 is designed for companies wishing to comply with ASTM D 1209, which specifies a Nessleriser system for colour comparison using 100 ml tubes, with the 100 ml graduation mark 275 to 295 mm above the bottom of the tube. Each system covers a set measuring range and includes.
  • A visual comparator instrument with a long path length, ensuring good sensitivity and accuracy when grading pale coloured samples
  • A selection of Pt-
More Product Information