Certified colour reference standards
Supplier: Lovibond Tintometer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
134000 134010 134050 134060 134200 134160 134190 109720 134210 134140 134150 134170 134180 133991
633-0004EA 207 GBP
633-0004 633-0005 633-0006 633-0007 633-0012 633-0056 633-0059 633-0258 633-0013 633-0008 633-0009 633-0010 633-0011 TINT133991
Certified colour reference standards
Standards Colour Reference and Spectroscopy Standards
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.