Magnesium sulphate 1.0 M MgSO4 volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

63126-1L
63126-1LEA 372 GBP
63126-1L
Magnesium sulphate 1.0 M MgSO4 volumetric solution, Fluka™
Magnesium sulphate
Formula: MgSO₄
Boiling Pt: 100 °C
Density: 1.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-791-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR