Staining jars acc. to Coplin
Supplier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42470010
631-9331EA 46.3 GBP
631-9331
Staining jars acc. to Coplin
Jars Staining Jars
Soda-lime glass.
For 10 standard microscopy slides (76×26 mm).