Staining jars
Supplier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42480010 42481010 42482010
631-9328EA 57.5 GBP
631-9328 631-9321 631-9329
Staining jars
Jars Staining Jars
Soda-lime glass.
  • For 10 standard slides  (76×26 mm)
Staining system contains staining jar, rack and handle made from stainless steel.
Ordering information: Insert and handle must be ordered separately.