Staining jars according to Coplin, WHEATON®

Supplier: DWK Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

900470
631-2508EA 288 GBP
631-2508
Staining jars according to Coplin, WHEATON®
Jars Staining Jars
Soda-lime glass.

  • Heavy glass walls and broad base for increased stability
  • Requires low reagent volume
  • Capacity: Approx. 65 ml

Staining jar for 5 single standard slides 76×26 mm vertically or 10 slides back to back with glass cover.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR