Staining dishes, WHEATON®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
900201 900200 900202 900203 900204 900205
631-2501EA 140 GBP
631-2501 631-2500 631-2502 631-2503 631-2504 631-2505
Staining dishes, WHEATON®
Jars Staining Jars
Soda-lime glass and stainless steel.
  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.