Microscope slides, UniMark®
Supplier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0704002 0704102 0704202 0704302 0704402 0704502 0703010 0703110 0703210 0703310 0703410 0703510
631-0093EA 9.7 GBP
631-0093 631-0094 631-0095 631-0096 631-0097 631-1209 631-1203 631-1204 631-1205 631-1206 631-1207 631-1208
Microscope slides, UniMark®
Slides Microscope Slides
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
  • Frosted end for labelling
  • Soda-lime glass, Hydrolytic Class 3
  • Different colours to identify preparations securely
  • Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.