VWR®, Slide Boxes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
631-0735EA 16.5 GBP
631-0735 631-0736 631-0737 631-0738 631-0739 631-0740
VWR®, Slide Boxes
Slide Boxes
Slide boxes for holding standard microscopy slides.
  • Hinged lids held securely by a latch lock
  • Stacking feet maximise stability when placed on top of one another
  • Slide inventory sheet on the inside of the lid
W×D×H: 90×140×35 mm (for 25), 165×210×35 mm (for 100)