Staining jars acc. to Schiefferdecker
Supplier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4200002
631-0699EA 293 GBP
631-0699
Staining jars acc. to Schiefferdecker
Jars Staining Jars
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.