VWR® Cover Glasses, Square
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
631-0124EA 65.1 GBP
631-0124 631-0121 631-0125 631-0120 631-0126 631-0127
VWR® Cover Glasses, Square
Cover Glasses
Made of pure white Hydrolytic Class 1 glass.
  • No bubbles or flaws
  • Cleaned and polished
Packaging: In a plastic box with hinged lid.
Order Now
SPECIFICATIONS
Environmentally Preferable