Reading magnifiers
Supplier: ESCHENBACH OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
264265 2642100
630-8150EA 62.2 GBP
630-8150 630-8151
Reading magnifiers
Magnifiers
This magnifier provides biconvex PXM® lightweight lens for an ergonomic use and optimised performances.
  • Additional plano-convex lens in the handle for 5× magnification
  • Eyelet in the handle, round-the-neck carry cord supplied
  • Antistatic coating