CELLCYTE X™ Live Cell Imaging and Analysis Inside Your Incubator
Supplier: DISCOVER ECHO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ECCX0200 ECCXT025 ECCXCW02
630-3520EA 63810 GBP
630-3520 630-3522 630-3521
CELLCYTE X™ Live Cell Imaging and Analysis Inside Your Incubator
Microscopes
CELLCYTE X™ offers the user the ability to image live cells in real-time from within the incubator. From all of these images, the user can fully understand and review cell kinetic trends at any point in time with user-friendly analysis software.
  • Built for successful experiments
  • Open design - Easier maintenance and environmental control
  • Real-time data analysis - Follow live cell kinetics as they occur
  • High throughput - Runs up to 6 vessels at the same time
  • Versatile - Ability to multiplex experiment with Enhanced Contour imaging mode plus 3 FL channels
  • High
More Product Information