VWR® VisiScope® 200, Upright Microscopes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
630-3291EA 538 GBP
630-3291 630-3292 630-3293 630-3294 630-3295
VWR® VisiScope® 200, Upright Microscopes
Microscopes
Reliable, advanced microscopes for educational purposes. Particularly recommended for secondary schools.
  • Powered illumination system with adjustable intensity – efficient LED energy saving lamp
  • Constant pure-white colour temperature at all intensity levels
  • Large opening behind the nosepiece for use as a handle
  • Optical components treated to prevent the formation of moulds and fungi
Certifications: Objectives conform to DIN standards. Strict controls on materials and production processes optimised by ISO 9001:2008 quality system.
Delivery information:More Product Information