Handheld digital microscopes, Dino-Lite Special Light, infrared (IR)
Supplier: Dino-Lite / IDCP B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AD4113T-I2V AM4115-FIT AM4115-FJT AM4115-FKT
IDCPAD4113T-I2VEA 655 GBP
IDCPAD4113T-I2V 630-2748 IDCPAM4115-FJT IDCPAM4115-FKT
Handheld digital microscopes, Dino-Lite Special Light, infrared (IR)
Microscopes
Handheld microscopes with infrared light or combinations between infrared and ultraviolet (UV) light.
  • Edge models provide superior image quality
  • MicroTouch trigger gives users the option of taking a picture right from the scope itself
  • High speed USB 2.0 output
  • All models (except AM7013M-FIT) feature exchangeable caps for more flexibility
  • Model AM7013M-FIT features an aluminium housing and is therefore ESD safe
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Models withMore Product Information