VWR® VisiCam® Plus, Digital USB Cameras
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
630-2737EA 392 GBP
630-2737 630-2738 630-2919 630-2973 630-2974 630-3357
VWR® VisiCam® Plus, Digital USB Cameras
Cameras
These cameras provide high resolution images for everyday microscopy.
  • Extremely reliable colour fidelity
  • Automatic exposure and white balance for clear images
  • USB 3.0 connection for high speed data transfer
  • Standard C-mount optical port
  • Includes software for previewing and capturing images and video for analysis and processing
With the correct C-mount for your microscope and camera, it fits to any microscope in educational, industrial or clinical use.
Accessories information: Required software can be downloaded online below "Related information".
More Product Information