Inverted fluorescence microscopes, Axio Vert A1 FL-LED
Supplier: ZEISS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4880140000000 4880499901400 4880019901000 0000001374896 4912370014000 4230529511000 4230529572000 4278099000000 4880509901000 4230529552000 4230529521000
ZEIS4880140000000EA 0 GBP
ZEIS4880140000000 ZEIS4880499901400 ZEIS4880019901000 ZEIS0000001374896 630-2152 ZEIS4230529511000 ZEIS4230529572000 ZEIS4278099000000 ZEIS4880509901000 ZEIS4230529552000 ZEIS4230529521000
Inverted fluorescence microscopes, Axio Vert A1 FL-LED
Microscopes
The Axio Vert A1 FL-LED has a four-position mount for LED modules for reflected light fluorescence. One position is for green fluorescent protein (GFP).
  • Differential interference contrast to observe the finest cells structures (optional)
  • LED fluorescence, long lifetime, homogeneous illumination with full intensity
  • No heating and cooling period required during the LED fluorescence
  • Ergonomic design to comfortably work in an upright position
  • Optional contrast systems (iHMC, PlasDIC, DIC)