Round cover glasses, Menzel Gläser
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CB00080RA120 CB00100RAC20 CB00220RAC20 CB00050RA120 CB00500RA140 CB00400RA140 CB00090RA120 CB00060RA120 CB00130RA120 CB00300RAC40 CB00320RA140 CB00120RAC20 CB00120RA020 CB00150RA220 CB00150RAC20 CB00190RA020 CB00190RA120 CB00190RA220 CB00220RA223 CB00250RA033 CB00350RA140 CB00190RAC20 CB00250RA233 CB00250RAC CB00180RAC20 CB00120RA2 CB00130RAC
MENZCB00080RA120EA 189 GBP
MENZCB00080RA120 630-2115 MENZCB00220RAC20 630-2112 630-2129 MENZCB00400RA140 MENZCB00090RA120 630-2113 630-2118 630-2124 630-2125 MENZCB00120RAC20 MENZCB00120RA020 MENZCB00150RA220 MENZCB00150RAC20 MENZCB00190RA020 MENZCB00190RA120 630-2120 MENZCB00220RA223 630-2122 630-2126 630-2121 630-2123 MENZCB00250RAC MENZCB00180RAC20 MENZCB00120RA2 MENZCB00130RAC
Round cover glasses, Menzel Gläser
Cover Glasses
The cover slips made of pure white glass of hydrolytic class 1.
Suitable for in vitro diagnostic (IVD) applications.