Adhesion slides, Menzel Gläser, SuperFrost® Plus
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
J1810AMNZ J1820AMNZ J1830AMNZ J1840AMNZ J1800AMNZ J7800AMNZ
630-0950EA 59.7 GBP
630-0950 630-0951 630-0952 630-0954 631-9483 MENZJ7800AMNZ
Adhesion slides, Menzel Gläser, SuperFrost® Plus
Slides Adhesive Slides
Superfrost® Plus slides hold tissue sections without the need for expensive coating steps.
  • Permanent positive charge, binds fresh, fixed or embedded tissue sections
  • Frosted ends for labelling
  • Made from fine sheet glass and selected for uniform quality
  • Annealed to minimise chipping and breakage
  • Resistant to corrosion or fogging
They eliminate tissue loss during staining, as well as hours of slide preparation by electrostatically attracting frozen tissue sections and cytology preparations, binding them to the slide.