VWR®, Camera Adapters
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
630-0865EA 55.3 GBP
630-0865 630-0866 630-0867 630-0868 630-2687
VWR®, Camera Adapters
Adapters Adapters, Camera to Microscope
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.